4-Chlorphenylsulfurpentafluorid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Chlorophenylsulphur Pentafluoride
PC5256-5G
APOSPC5256-5GEA
0
EUR
APOSPC5256-5G
4-Chlorphenylsulfurpentafluorid
4-Chlorphenylsulfurpentafluorid
Formel:
C₆H₄ClF₅S Molecular Weight: 238.606 g/mol Siedepunkt: 78 °C/9.7mmHg Flammpunkt: 75 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD11501051 CAS-Nummer: 5310-68-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...