2-Nitro-6-(trifluormethyl)anisol
Lieferant: Apollo Scientific
|
Gefahr
|
PC5267-1G
PC5267-5G
APOSPC5267-1GEA
0
EUR
APOSPC5267-1G
APOSPC5267-5G
2-Nitro-6-(trifluormethyl)anisol
2-Nitro-6-(trifluormethyl)anisol
Formel:
C₈H₆F₃NO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD08532488 CAS-Nummer: 627531-40-2 UN: 1760 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...