2-Nitro-6-(trifluormethyl)anisol

Lieferant: Apollo Scientific
Gefahr

PC5267-1G PC5267-5G
APOSPC5267-1GEA 0 EUR
APOSPC5267-1G APOSPC5267-5G
2-Nitro-6-(trifluormethyl)anisol
2-Nitro-6-(trifluormethyl)anisol
Formel: C₈H₆F₃NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD08532488
CAS-Nummer: 627531-40-2
UN: 1760
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR