4-Bromo-2-chloro-3-fluorobenzaldehyde 95%

Lieferant: Apollo Scientific

PC52714-1G PC52714-250MG PC52714-5G
APOSPC52714-1GEA 543 EUR
APOSPC52714-1G APOSPC52714-250MG APOSPC52714-5G
4-Bromo-2-chloro-3-fluorobenzaldehyde 95%
4-Bromo-2-chloro-3-fluorobenzaldehyde
Formel: C₇H₃BrClFO
Molecular Weight: 237.454 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28733869
CAS-Nummer: 1780699-03-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR