2-Nitro-4-(trifluormethoxy)anilin 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC5292-25G
PC5292-5G
PC5292-100G
APOSPC5292-25GEA
26.1
EUR
APOSPC5292-25G
APOSPC5292-5G
APOSPC5292-100G
2-Nitro-4-(trifluormethoxy)anilin 98%
2-Nitro-4-(trifluormethoxy)anilin
Formel:
C₇H₅F₃N₂O₃ Schmelzpunkt: 62…64 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00042326 CAS-Nummer: 2267-23-4 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...