2,5-Difluoro-L-phenylalanine 97%

Lieferant: Apollo Scientific

Synonyme: (S)-2-Amino-3-(2,5-difluorophenyl)propanoic acid, H-Phe(2,5-DiF)-OH

PC530006-1G
APOSPC530006-1GEA 116 EUR
APOSPC530006-1G
2,5-Difluoro-L-phenylalanine 97%
2,5-Difluoro-L-phenylalanine
Formel: C₉H₉F₂NO₂
Molecular Weight: 201.17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07372000
CAS-Nummer: 31105-92-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR