3,4,5-Trifluoro-L-phenylalanine 97%

Lieferant: Apollo Scientific

Synonyme: (S)-2-Amino-3-(3,4,5-trifluorophenyl)propionic acid, H-Phe(3,4,5-TriF)-OH

PC530007-1G
APOSPC530007-1GEA 290 EUR
APOSPC530007-1G
3,4,5-Trifluoro-L-phenylalanine 97%
3,4,5-Trifluoro-L-phenylalanine
Formel: C₉H₈F₃NO₂
Molecular Weight: 219.161 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03428422
CAS-Nummer: 646066-73-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR