4-Fluoro-DL-homophenylalanine 97%

Lieferant: Apollo Scientific

PC530008-1G PC530008-250MG PC530008-500MG
APOSPC530008-1GEA 654 EUR
APOSPC530008-1G APOSPC530008-250MG APOSPC530008-500MG
4-Fluoro-DL-homophenylalanine 97%
4-Fluoro-DL-homophenylalanine
Formel: C₁₀H₁₂FNO₂
Molecular Weight: 197.206 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07372156
CAS-Nummer: 225233-79-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR