N-Boc-(S)-2,4,5-trifluoro-b-homophenylalanine 97%

Lieferant: Apollo Scientific

Synonyme: (S)-3-((tert-Butoxycarbonyl)amino)-4-(2,4,5-trifluorophenyl)butanoic acid

PC530015-1G
APOSPC530015-1GEA 571 EUR
APOSPC530015-1G
N-Boc-(S)-2,4,5-trifluoro-b-homophenylalanine 97%
N-Boc-(S)-2,4,5-trifluoro-b-homophenylalanine
Formel: C₁₅H₁₈F₃NO₄
Molecular Weight: 333.303 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06659150
CAS-Nummer: 922178-94-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR