1-(3-Fluorphenyl)cyclopentancarbonsäure 97%

Lieferant: Apollo Scientific

PC530022-5G
APOSPC530022-5GEA 161 EUR
APOSPC530022-5G
1-(3-Fluorphenyl)cyclopentancarbonsäure 97%
1-(3-Fluorophenyl)cyclopentanecarboxylic acid
Formel: C₁₂H₁₃FO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00800624
CAS-Nummer: 214262-97-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR