3-(Trifluoromethyl)pyrrolidine-3-ol hydrochloride 97%

Lieferant: Apollo Scientific

Synonyme: 3-(Trifluoromethyl)-3-pyrrolidinol hydrochloride, 3-(Trifluoromethyl)pyrrolidin-3-ol hydrochloride

PC530026-1G
APOSPC530026-1GEA 207 EUR
APOSPC530026-1G
3-(Trifluoromethyl)pyrrolidine-3-ol hydrochloride 97%
3-(Trifluoromethyl)pyrrolidine-3-ol hydrochloride
Formel: C₅H₉ClF₃NO
Molecular Weight: 191.58 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19982573
CAS-Nummer: 1334147-81-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR