4-(2-Fluorobenzyl)piperidine hydrochloride 97%

Lieferant: Apollo Scientific

PC530027-1G
APOSPC530027-1GEA 116 EUR
APOSPC530027-1G
4-(2-Fluorobenzyl)piperidine hydrochloride 97%
4-(2-Fluorobenzyl)piperidine hydrochloride
Formel: C₁₂H₁₆FN·ClH
Molecular Weight: 229,721 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09787475
CAS-Nummer: 193357-26-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR