Boc-5-Fluoro-D-tryptophan 97%

Lieferant: Apollo Scientific

PC530030-1G PC530030-250MG PC530030-500MG
APOSPC530030-1GEA 0 EUR
APOSPC530030-1G APOSPC530030-250MG APOSPC530030-500MG
Boc-5-Fluoro-D-tryptophan 97%
Boc-5-Fluoro-D-tryptophan
Formel: C₁₆H₁₉FN₂O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD09264332
CAS-Nummer: 114926-41-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR