6-Fluoro-L-tryptophan 97%

Lieferant: Apollo Scientific

PC530036-1G PC530036-250MG
APOSPC530036-1GEA 0 EUR
APOSPC530036-1G APOSPC530036-250MG
6-Fluoro-L-tryptophan 97%
6-Fluoro-L-tryptophan
Formel: C₁₁H₁₁FN₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00137731
CAS-Nummer: 19310-00-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR