2-Amino-5-iodo-4-(trifluoromethyl)-1,3-thiazole 95+%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Iodo-4-(trifluoromethyl)-1,3-thiazol-2-amine, 5-Iodo-4-(trifluoromethyl)thiazol-2-amine
PC53004-250MG
APOSPC53004-250MGEA
197
EUR
APOSPC53004-250MG
2-Amino-5-iodo-4-(trifluoromethyl)-1,3-thiazole 95+%
2-Amino-5-iodo-4-(trifluoromethyl)-1,3-thiazole
Formel:
C₄H₂F₃IN₂S Molecular Weight: 294,037 g/mol Schmelzpunkt: 108…109 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD11847617 CAS-Nummer: 682342-65-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...