2-Amino-1-(2-fluorophenyl)ethanol 97%

Lieferant: Apollo Scientific

Synonyme: 2-Amino-1-(2-fluorophenyl)ethanol hydrochloride

PC530041-1G PC530041-500MG
APOSPC530041-1GEA 654 EUR
APOSPC530041-1G APOSPC530041-500MG
2-Amino-1-(2-fluorophenyl)ethanol 97%
2-Amino-1-(2-fluorophenyl)ethanol
Formel: C₈H₁₀FNO
Lagertemperatur: Raumtemperatur
MDL: MFCD08060976
CAS-Nummer: 3225-74-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR