3-Bromo-2-chloro-6-fluorobenzoic acid

Lieferant: Apollo Scientific
Achtung

PC53025-1G PC53025-5G
APOSPC53025-1GEA 65.4 EUR
APOSPC53025-1G APOSPC53025-5G
3-Bromo-2-chloro-6-fluorobenzoic acid
3-Bromo-2-chloro-6-fluorobenzoic acid
Formel: C₇H₃BrClFO₂
Schmelzpunkt: 134…136 °C
Lagertemperatur: Kühlschrank
MDL: MFCD11856020
CAS-Nummer: 1114809-13-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR