5-Bromo-6-fluoroisatoic anhydride

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Bromo-5-fluoro-2H-3,1-benzoxazine-2,4(1H)-dione

PC53036-1G PC53036-5G
APOSPC53036-1GEA 447 EUR
APOSPC53036-1G APOSPC53036-5G
5-Bromo-6-fluoroisatoic anhydride
5-Bromo-6-fluoroisatoic anhydride
Formel: C₈H₃BrFNO₃
Molecular Weight: 260.017 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28897196
CAS-Nummer: 1935441-96-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR