5-Fluoro-2-methoxy-4-nitroaniline 95+%

Lieferant: Apollo Scientific
Achtung

PC53063-1G
APOSPC53063-1GEA 101 EUR
APOSPC53063-1G
5-Fluoro-2-methoxy-4-nitroaniline 95+%
5-Fluoro-2-methoxy-4-nitroaniline
Formel: C₇H₇FN₂O₃
Molecular Weight: 186,14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24842657
CAS-Nummer: 1435806-78-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR