Potassium benzyl N-[2-(trifluoroboranuidyl)ethyl]carbamate 95%

Lieferant: Apollo Scientific

Synonyme: Potassium (2-(((benzyloxy)carbonyl)amino)ethyl)trifluoroborate, Potassium (2-{[(benzyloxy)carbonyl]amino}ethyl)(trifluoro)borate

PC53080-1G
APOSPC53080-1GEA 116 EUR
APOSPC53080-1G
Potassium benzyl N-[2-(trifluoroboranuidyl)ethyl]carbamate 95%
Potassium benzyl N-[2-(trifluoroboranuidyl)ethyl]carbamate
Formel: C₁₀H₁₂BF₃NO₂·K
Molecular Weight: 285,112 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10698612
CAS-Nummer: 926280-84-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR