Perfluorobutyl bromide 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC5309-5G
PC5309-1G
APOSPC5309-5GEA
0
EUR
APOSPC5309-5G
APOSPC5309-1G
Perfluorobutyl bromide 97%
Perfluorobutyl bromide
Formel:
C₄BrF₉ Siedepunkt: 43…44 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00082622 CAS-Nummer: 375-48-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...