(S)-1-(4-(Trifluoromethoxy)phenyl)ethanamine hydrochloride 95+%

Lieferant: Apollo Scientific

PC53093-250MG
APOSPC53093-250MGEA 162 EUR
APOSPC53093-250MG
(S)-1-(4-(Trifluoromethoxy)phenyl)ethanamine hydrochloride 95+%
(S)-1-(4-(Trifluoromethoxy)phenyl)ethanamine hydrochloride
Formel: C₉H₁₀F₃NO·ClH
Molecular Weight: 241.638 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12757222
CAS-Nummer: 1391540-47-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR