3-Chloro-3,3-difluoroprop-1-ene 99%

Lieferant: Apollo Scientific
Achtung

Synonyme: HFO-1242zf, 3-Difluoro-3-chloro-1-propene

PC53102-5G
APOSPC53102-5GEA 638 EUR
APOSPC53102-5G
3-Chloro-3,3-difluoroprop-1-ene 99%
3-Chloro-3,3-difluoroprop-1-ene
Formel: C₃H₃ClF₂
Molecular Weight: 112,506 g/mol
Siedepunkt: 17…18 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD28987627
CAS-Nummer: 421-03-4
UN: 3161
ADR: 2.1,

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR