2-Bromo-4-fluoro-5-nitrotoluene 98%

Lieferant: Apollo Scientific

Synonyme: 1-Bromo-5-fluoro-2-methyl-4-nitrobenzene

PC53113-5G
APOSPC53113-5GEA 53.8 EUR
APOSPC53113-5G
2-Bromo-4-fluoro-5-nitrotoluene 98%
2-Bromo-4-fluoro-5-nitrotoluene
Formel: C₇H₅BrFNO₂
Molecular Weight: 234,023 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11847636
CAS-Nummer: 64695-96-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR