1-(4-Fluoro-3-nitrophenyl)-4-methylpiperazine

Lieferant: Apollo Scientific

PC53116-250MG
APOSPC53116-250MGEA 713 EUR
APOSPC53116-250MG
1-(4-Fluoro-3-nitrophenyl)-4-methylpiperazine
1-(4-Fluoro-3-nitrophenyl)-4-methylpiperazine
Formel: C₁₁H₁₄FN₃O₂
Molecular Weight: 239,246 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD15142588
CAS-Nummer: 1216614-11-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR