(S)-1-(6-(Trifluoromethyl)-4-methylpyridin-3-yl)ethanamine

Lieferant: Apollo Scientific

PC53141-500MG
APOSPC53141-500MGEA 466 EUR
APOSPC53141-500MG
(S)-1-(6-(Trifluoromethyl)-4-methylpyridin-3-yl)ethanamine
(S)-1-(6-(Trifluoromethyl)-4-methylpyridin-3-yl)ethanamine
Formel: C₉H₁₁F₃N₂
Molecular Weight: 204,192 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29042490
CAS-Nummer: 1932509-91-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR