(S)-1-(6-(Trifluoromethyl)pyridin-3-yl)ethanamine
Lieferant: Apollo Scientific
|
Achtung
|
PC53142-100MG
APOSPC53142-100MGEA
278
EUR
APOSPC53142-100MG
(S)-1-(6-(Trifluoromethyl)pyridin-3-yl)ethanamine
(S)-1-(6-(Trifluoromethyl)pyridin-3-yl)ethanamine
Formel:
C₈H₉F₃N₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD09256747 CAS-Nummer: 1071435-62-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...