(S)-1-(3-Bromo-5-fluoropyridin-2-yl)ethanamine
Lieferant: Apollo Scientific
PC53143-500MG
APOSPC53143-500MGEA
466
EUR
APOSPC53143-500MG
(S)-1-(3-Bromo-5-fluoropyridin-2-yl)ethanamine
(S)-1-(3-Bromo-5-fluoropyridin-2-yl)ethanamine
Formel:
C₇H₈BrFN₂ Molecular Weight: 219,054 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD29042491 CAS-Nummer: 1932587-71-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...