(S)-1-(5-Chloro-3-fluoropyridin-2-yl)ethanamine

Lieferant: Apollo Scientific

PC53144-500MG
APOSPC53144-500MGEA 466 EUR
APOSPC53144-500MG
(S)-1-(5-Chloro-3-fluoropyridin-2-yl)ethanamine
(S)-1-(5-Chloro-3-fluoropyridin-2-yl)ethanamine
Formel: C₇H₈ClFN₂
Molecular Weight: 174,603 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29042492
CAS-Nummer: 1932020-62-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR