3-Bromo-2-(trifluoromethyl)aniline

Lieferant: Apollo Scientific
Achtung

PC53158-1G
APOSPC53158-1GEA 273 EUR
APOSPC53158-1G
3-Bromo-2-(trifluoromethyl)aniline
3-Bromo-2-(trifluoromethyl)aniline
Formel: C₇H₅BrF₃N
Lagertemperatur: Kühlschrank
MDL: MFCD13194482
CAS-Nummer: 244246-71-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR