6-Fluoropyrazin-2-amine
Lieferant: Apollo Scientific
|
Gefahr
|
PC53159-1G
APOSPC53159-1GEA
546
EUR
APOSPC53159-1G
6-Fluoropyrazin-2-amine
6-Fluoropyrazin-2-amine
Formel:
C₄H₄FN₃ Molecular Weight: 113,093 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD22566352 CAS-Nummer: 1246466-74-9 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...