3-Fluoropyrazin-2-amine
Lieferant: Apollo Scientific
|
Gefahr
|
PC53161-1G
APOSPC53161-1GEA
546
EUR
APOSPC53161-1G
3-Fluoropyrazin-2-amine
3-Fluoropyrazin-2-amine
Formel:
C₄H₄FN₃ Molecular Weight: 113.093 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD23382413 CAS-Nummer: 1206523-95-6 UN: 1993 ADR: 3 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...