5-Oxo-5-(3-trifluoromethylphenyl)valeric acid

Lieferant: Apollo Scientific
Achtung

PC53173-2G
APOSPC53173-2GEA 1410 EUR
APOSPC53173-2G
5-Oxo-5-(3-trifluoromethylphenyl)valeric acid
5-Oxo-5-(3-trifluoromethylphenyl)valeric acid
Formel: C₁₂H₁₁F₃O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD02260917
CAS-Nummer: 502650-98-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR