2-Methyl-7-(trifluoromethyl)quinoline 95+%

Lieferant: Apollo Scientific
Achtung

PC53178-250MG
APOSPC53178-250MGEA 128 EUR
APOSPC53178-250MG
2-Methyl-7-(trifluoromethyl)quinoline 95+%
2-Methyl-7-(trifluoromethyl)quinoline
Formel: C₁₁H₈F₃N
Molecular Weight: 211,183 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD11108688
CAS-Nummer: 324-32-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR