2-[4-(4-Fluoro-2-nitrophenyl)piperazin-1-yl]ethanol hydrochloride

Lieferant: Apollo Scientific

PC5318-1G
APOSPC5318-1GEA 184 EUR
APOSPC5318-1G
2-[4-(4-Fluoro-2-nitrophenyl)piperazin-1-yl]ethanol hydrochloride
2-[4-(4-Fluoro-2-nitrophenyl)piperazin-1-yl]ethanol hydrochloride
Formel: C₁₂H₁₇ClFN₃O₃
Schmelzpunkt: 220…223 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08692462
CAS-Nummer: 1185297-30-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR