2,6-Difluoro-3,5-dinitrotoluene

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,4-Difluoro-3-methyl-1,5-dinitrobenzene

PC53183-1G PC53183-250MG
APOSPC53183-1GEA 509 EUR
APOSPC53183-1G APOSPC53183-250MG
2,6-Difluoro-3,5-dinitrotoluene
2,6-Difluoro-3,5-dinitrotoluene
Formel: C₇H₄F₂N₂O₄
Lagertemperatur: Kühlschrank
MDL: MFCD16620286
CAS-Nummer: 112822-76-1
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR