[3-(Trifluoromethoxy)phenyl]hydrazine hydrochloride
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(3-(Trifluoromethoxy)phenyl)hydrazine hydrochloride
PC53187-25G
APOSPC53187-25GEA
155
EUR
APOSPC53187-25G
[3-(Trifluoromethoxy)phenyl]hydrazine hydrochloride
[3-(Trifluoromethoxy)phenyl]hydrazine hydrochloride
Formel:
C₇H₇F₃N₂O·ClH Molecular Weight: 228.599 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD02671043 CAS-Nummer: 133115-55-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...