1-(2-Fluor-4-nitrophenyl)piperidin-4-carbonsäureamid

Lieferant: Apollo Scientific

PC5319-1G
APOSPC5319-1GEA 255 EUR
APOSPC5319-1G
1-(2-Fluor-4-nitrophenyl)piperidin-4-carbonsäureamid
1-(2-Fluor-4-nitrophenyl)piperidin-4-carbonsäureamid
Formel: C₁₂H₁₄FN₃O₃
Schmelzpunkt: 226…227 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08692380
CAS-Nummer: 912894-29-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR