6-Fluoro-1,2,3,4-tetrahydroisoquinoline hydrochloride 95%

Lieferant: Apollo Scientific
Achtung

PC53191-250MG
APOSPC53191-250MGEA 98.6 EUR
APOSPC53191-250MG
6-Fluoro-1,2,3,4-tetrahydroisoquinoline hydrochloride 95%
6-Fluoro-1,2,3,4-tetrahydroisoquinoline hydrochloride
Formel: C₉H₁₀FN·ClH
Molecular Weight: 187,642 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08437639
CAS-Nummer: 799274-08-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR