5-Bromo-6-fluoro-1H-indole 96%

Lieferant: Apollo Scientific
Achtung

PC53195-250MG
APOSPC53195-250MGEA 195 EUR
APOSPC53195-250MG
5-Bromo-6-fluoro-1H-indole 96%
5-Bromo-6-fluoro-1H-indole
Formel: C₈H₅BrFN
Molecular Weight: 214,034 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD11848565
CAS-Nummer: 434960-42-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR