3-Bromo-2-fluoropyridine-5-boronic acid, pinacol ester 98%

Lieferant: Apollo Scientific
Achtung

PC53200-1G
APOSPC53200-1GEA 664 EUR
APOSPC53200-1G
3-Bromo-2-fluoropyridine-5-boronic acid, pinacol ester 98%
3-Bromo-2-fluoropyridine-5-boronic acid, pinacol ester
Formel: C₁₁H₁₄BBrFNO₂
Lagertemperatur: Kühlschrank
MDL: MFCD16995269
CAS-Nummer: 2223038-87-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR