2,3-Difluor-5-hydroxybenzoesäure
Lieferant: Apollo Scientific
|
Achtung
|
PC53217-1G
APOSPC53217-1GEA
473
EUR
APOSPC53217-1G
2,3-Difluor-5-hydroxybenzoesäure
2,3-Difluoro-5-hydroxybenzoic acid
Formel:
C₇H₄F₂O₃ Schmelzpunkt: 180…185 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08704572 CAS-Nummer: 749230-51-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...