1-(4-Fluor-2-nitrophenyl)piperidin-4-carbonsäureamid
Lieferant: Apollo Scientific
PC5322-1G
APOSPC5322-1GEA
184
EUR
APOSPC5322-1G
1-(4-Fluor-2-nitrophenyl)piperidin-4-carbonsäureamid
1-(4-Fluor-2-nitrophenyl)piperidin-4-carbonsäureamid
Formel:
C₁₂H₁₄FN₃O₃ Schmelzpunkt: 163…164 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD05664788 CAS-Nummer: 874800-67-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...