Potassium (E)-propenyl-1-trifluoroborate

Lieferant: Apollo Scientific
Gefahr

Synonyme: Potassium (E)-trifluoro(prop-1-en-1-yl)borate

PC53224-1G
APOSPC53224-1GEA 139 EUR
APOSPC53224-1G
Potassium (E)-propenyl-1-trifluoroborate
Potassium (E)-propenyl-1-trifluoroborate
Formel: C₃H₅BF₃·K
Molecular Weight: 147,976 g/mol
Schmelzpunkt: 81 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09992882
CAS-Nummer: 1902198-18-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR