1,1,1-Trifluoro-2-(piperidin-4-yl)propan-2-ol
Lieferant: Apollo Scientific
PC53225-250MG
APOSPC53225-250MGEA
851
EUR
APOSPC53225-250MG
1,1,1-Trifluoro-2-(piperidin-4-yl)propan-2-ol
1,1,1-Trifluoro-2-(piperidin-4-yl)propan-2-ol
Formel:
C₈H₁₄F₃NO Lagertemperatur: Raumtemperatur |
MDL:
MFCD17977193 CAS-Nummer: 1227068-37-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...