5-Bromo-6-fluoropyridine-3-carbonitrile 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Bromo-6-fluoronicotinonitrile

PC53229-250MG
APOSPC53229-250MGEA 480 EUR
APOSPC53229-250MG
5-Bromo-6-fluoropyridine-3-carbonitrile 97%
5-Bromo-6-fluoropyridine-3-carbonitrile
Formel: C₆H₂BrFN₂
Molecular Weight: 200,996 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18258641
CAS-Nummer: 1256823-71-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR