1-(2-Fluor-4-nitrophenyl)piperidin-2-carbonsäure

Lieferant: Apollo Scientific

PC5323-500MG
APOSPC5323-500MGEA 390 EUR
APOSPC5323-500MG
1-(2-Fluor-4-nitrophenyl)piperidin-2-carbonsäure
1-(2-Fluor-4-nitrophenyl)piperidin-2-carbonsäure
Formel: C₁₂H₁₃FN₂O₄
Schmelzpunkt: 153…154 °C
Lagertemperatur: Raumtemperatur
CAS-Nummer: 1214189-74-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR