1-Boc-3-fluoropyrrolidine-3-methylamine

Lieferant: Apollo Scientific
Achtung

Synonyme: tert-Butyl 3-(aminomethyl)-3-fluoropyrrolidine-1-carboxylate

PC53232-1G
APOSPC53232-1GEA 1090 EUR
APOSPC53232-1G
1-Boc-3-fluoropyrrolidine-3-methylamine
1-Boc-3-fluoropyrrolidine-3-methylamine
Formel: C₁₀H₁₉FN₂O₂
Molecular Weight: 218,27 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18375054
CAS-Nummer: 1228765-06-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR