1-(2-Fluor-4-nitrophenyl)piperidin-4-carbonsäure

Lieferant: Apollo Scientific

PC5324-1G
APOSPC5324-1GEA 326 EUR
APOSPC5324-1G
1-(2-Fluor-4-nitrophenyl)piperidin-4-carbonsäure
1-(2-Fluor-4-nitrophenyl)piperidin-4-carbonsäure
Formel: C₁₂H₁₃FN₂O₄
Schmelzpunkt: 187…188 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08692407
CAS-Nummer: 325701-99-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR