(2,4-Difluorphenyl)aceton 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-(2,4-Difluorophenyl)propan-2-one
PC53241-1G
PC53241-250MG
PC53241-5G
APOSPC53241-1GEA
166
EUR
APOSPC53241-1G
APOSPC53241-250MG
APOSPC53241-5G
(2,4-Difluorphenyl)aceton 97%
(2,4-Difluorphenyl)aceton
Formel:
C₉H₈F₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD02258866 CAS-Nummer: 274682-91-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...